missing translation for 'onlineSavingsMsg'
Learn More

Homotaurine, TRC

Product Code. 30398494
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30398494 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398494 Supplier Toronto Research Chemicals Supplier No. TRCH65620050MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Aminopropane-1-sulfonic Acid (Homotaurine)
CAS 3687-18-1
Molecular Formula C3 H9 N O3 S
Quantity 50 mg
Synonym 3-Aminopropane-1-sulfonic Acid, Homotaurine, 1-Propanesulfonic acid, 3-amino- (6CI, 7CI, 8CI, 9CI, ACI), 3-Amino-1-propanesulfonic acid (ACI), 3-Amino-1-propanesulfonate, 3-Aminopropanesulfonic acid, 3-Aminopropiosulfonic acid, 3-Azaniumylpropane-1-sulfonate, 3-Sulfopropylamine, Alzhemed, NC 758, NSC 77071, Tramiprosate, γ-Aminopropylsulfonic acid
InChI Formula InChI=1S/C3H9NO3S/c4-2-1-3-8(5,6)7/h1-4H2,(H,5,6,7)
SMILES NCCCS(=O)(=O)O
IUPAC Name 3-aminopropane-1-sulfonic acid
Molecular Weight (g/mol) 139.17
Formula Weight 139.0303
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.