missing translation for 'onlineSavingsMsg'
Learn More

Homosibutramine, TRC

Product Code. 30401612
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401612 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401612 Supplier Toronto Research Chemicals Supplier No. TRCH6151535MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Homosibutramine
CAS 935888-80-5
Molecular Formula C18 H28 Cl N
Quantity 5 mg
Synonym Homo Sibutramine
InChI Formula InChI=1S/C18H28ClN/c1-5-20(4)17(13-14(2)3)18(11-6-12-18)15-7-9-16(19)10-8-15/h7-10,14,17H,5-6,11-13H2,1-4H3
SMILES CCN(C)C(CC(C)C)C1(CCC1)c2ccc(Cl)cc2
IUPAC Name 1-[1-(4-chlorophenyl)cyclobutyl]-N-ethyl-N,3-dimethylbutan-1-amine
Molecular Weight (g/mol) 293.875
Formula Weight 293.191
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.