missing translation for 'onlineSavingsMsg'
Learn More

Homopiperazine-N,N’-bis-[2-(ethanesulfonic acid)] Disodium Salt, TRC

Product Code. 30408524
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
This item is not returnable. View return policy

Product Code. 30408524

Brand: Toronto Research Chemicals TRCH6015005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Homopiperazine-N,N’-bis-[2-(ethanesulfonic acid)] Disodium Salt
Molecular Formula C9 H18 N2 Na2 O6 S2
Quantity 5 g
Synonym Tetrahydro-1H-1,4-diazepine-1,4(5H)-diethanesulfonic Acid Disodium Salt
InChI Formula InChI=1S/C9H20N2O6S2.2Na/c12-18(13,14)8-6-10-2-1-3-11(5-4-10)7-9-19(15,16)17;;/h1-9H2,(H,12,13,14)(H,15,16,17);;/q;2*+1/p-2
SMILES [Na]OS(=O)(=O)CCN1CCCN(CCS(=O)(=O)O[Na])CC1
IUPAC Name 2-[4-(2-sodiooxysulfonylethyl)-1,4-diazepan-1-yl]ethylsulfonyloxysodium
Molecular Weight (g/mol) 360.359
Formula Weight 360.04
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.