missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homopiperazine-N,N’-bis-[2-(ethanesulfonic acid)] Disodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Homopiperazine-N,N’-bis-[2-(ethanesulfonic acid)] Disodium Salt |
| Molecular Formula | C9 H18 N2 Na2 O6 S2 |
| Quantity | 10 g |
| Synonym | Tetrahydro-1H-1,4-diazepine-1,4(5H)-diethanesulfonic Acid Disodium Salt |
| InChI Formula | InChI=1S/C9H20N2O6S2.2Na/c12-18(13,14)8-6-10-2-1-3-11(5-4-10)7-9-19(15,16)17;;/h1-9H2,(H,12,13,14)(H,15,16,17);;/q;2*+1/p-2 |
| SMILES | [Na]OS(=O)(=O)CCN1CCCN(CCS(=O)(=O)O[Na])CC1 |
| IUPAC Name | 2-[4-(2-sodiooxysulfonylethyl)-1,4-diazepan-1-yl]ethylsulfonyloxysodium |
| Molecular Weight (g/mol) | 360.359 |
| Formula Weight | 360.04 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?