missing translation for 'onlineSavingsMsg'
Learn More

Homophthalic anhydride, 97+%, Thermo Scientific Chemicals

Product Code. 11462967
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462967 50 g 50g
11452967 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11462967 Supplier Thermo Scientific Alfa Aesar Supplier No. A15538.18
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 703-59-3 | C9H6O3 | 162.144 g/mol

It is used as a chemical and organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a chemical and organic intermediates.

Solubility
Decomposes in water.

Notes
Moisture sensitive. Storage temperature 15 - 30°C. Store under inert gas. Store away from exposure to moisture, strong oxidizing agents, strong acids, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 703-59-3
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.144
MDL Number MFCD00006894
InChI Key AKHSBAVQPIRVAG-UHFFFAOYSA-N
Synonym homophthalic anhydride, 1,3-isochromandione, isochroman-1,3-dione, 1h-2-benzopyran-1,3 4h-dione, homophthalic acid anhydride, 4h-2-benzopyran-1,3-dione, 1h-isochromene-1,3 4h-dione, 3,4-dihydro-1h-2-benzopyran-1,3-dione, 4h-benzo c pyran-1,3-dione, homophthalic anhydride 2-carboxyphenylacetic anhydride
PubChem CID 12801
IUPAC Name 4H-isochromene-1,3-dione
SMILES C1C2=CC=CC=C2C(=O)OC1=O

Specifications

Melting Point 140°C to 144°C
Quantity 50 g
Beilstein 5244
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 162.14
Percent Purity ≥97%
Chemical Name or Material Homophthalic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.