missing translation for 'onlineSavingsMsg'
Learn More

Homophthalic acid, 99%

Product Code. 11402477
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402477 50 g 50g
11412477 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11402477 Supplier Thermo Scientific Alfa Aesar Supplier No. A14297.18
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 89-51-0 | C9H8O4 | 180.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-51-0
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00004326
InChI Key ZHQLTKAVLJKSKR-UHFFFAOYSA-N
Synonym homophthalic acid, 2-carboxymethyl benzoic acid, benzeneacetic acid, 2-carboxy, 2-carboxyphenylacetic acid, 2-carboxybenzeneacetic acid, alpha-carboxy-o-toluic acid, o-carboxymethyl benzoic acid, 2-carboxyphenyl acetic acid, o-toluic acid, alpha-carboxy, toluene-alpha,2-dicarboxylic acid
PubChem CID 66643
IUPAC Name 2-(carboxymethyl)benzoic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)C(=O)O

Specifications

Melting Point ∼180°C (decomposition)
Quantity 50 g
Beilstein 1872069
Formula Weight 180.16
Percent Purity 99%
Chemical Name or Material Homophthalic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.