missing translation for 'onlineSavingsMsg'
Learn More

Homogentisic acid, MedChemExpress

Product Code. 16454768
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16454768 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16454768 Supplier Medchem Express Supplier No. HY113283
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Homogentisic acid is a specific metabolite in urine and serum, which is used for diagnosis of alkaptonuria.

Homogentisic acid is a specific metabolite in urine and serum, which is used for diagnosis of alkaptonuria.

Specifications

Chemical Name or Material Homogentisic acid
CAS 451-13-8
Color White
Purity Grade Notes Research
Molecular Formula C8H8O4
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (594.71 mM; Need ultrasonic)
SMILES O=C(O)CC1=CC(O)=CC=C1O
Molecular Weight (g/mol) 168.15
Formula Weight 168.15
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.