missing translation for 'onlineSavingsMsg'
Learn More

Homocapsaicin I, TRC

Product Code. 30391663
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391663 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391663 Supplier Toronto Research Chemicals Supplier No. TRCH58990010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Homocapsaicin I
CAS 58493-48-4
Molecular Formula C19 H29 N O3
Quantity 10 mg
Synonym Homocapsaicin I, (E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-9-methyl-7-Decenamide, (7E)-N-[(4-Hydroxy-3-methoxyphenyl)methyl]-9-methyl-7-decenamide, E-Homocapsaicin, Homocapsaicin
InChI Formula InChI=1S/C19H29NO3/c1-15(2)9-7-5-4-6-8-10-19(22)20-14-16-11-12-17(21)18(13-16)23-3/h7,9,11-13,15,21H,4-6,8,10,14H2,1-3H3,(H,20,22)/b9-7+
SMILES COc1cc(CNC(=O)CCCCC\C=C\C(C)C)ccc1O
IUPAC Name (E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-9-methyldec-7-enamide
Molecular Weight (g/mol) 319.438
Formula Weight 319.215
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.