missing translation for 'onlineSavingsMsg'
Learn More

Homoallylglycine, TRC

Product Code. 30396063
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396063 50 mg
30401098 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396063 Supplier Toronto Research Chemicals Supplier No. TRCH58401050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16258-05-2
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
Synonym 2-Amino-5-hexenoic Acid
IUPAC Name 2-aminohex-5-enoic acid
SMILES NC(CCC=C)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H11NO2/c1-2-3-4-5(7)6(8)9/h2,5H,1,3-4,7H2,(H,8,9)
Formula Weight 129.079
Chemical Name or Material Homoallylglycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.