missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homez Mouse anti-Human, Clone: PCRP-HOMEZ-1B5, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | Homez |
|---|---|
| Clone | PCRP-HOMEZ-1B5 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Peptide Array |
Description
Homez Monoclonal specifically detects Homez in Human samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Homez | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| homeobox and leucine zipper encoding, homeobox and leucine zipper protein Homez, KIAA1443Homeodomain leucine zipper-containing factor | |
| Recombinant full-length human Homez protein | |
| Primary | |
| Store at 4°C. |
| PCRP-HOMEZ-1B5 | |
| Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 57594 | |
| IgG2a | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title