missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOMER2 Antibody (OTI1H3), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71465APC
This item is not returnable.
View return policy
Description
HOMER2 Monoclonal antibody specifically detects HOMER2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| HOMER2 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI1H3 | |
| Western Blot | |
| ACPD, CPD, cupidin, homer homolog 2 (Drosophila), homer homolog 3, homer protein homolog 2, homer, neuronal immediate early gene, 2, Homer-2, HOMER2A, HOMER-2A, HOMER2B, HOMER-2B, Vesl-2 | |
| Full length human recombinant protein of human HOMER2(NP_004830) produced in E.coli. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 9455 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction