missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HOMER1 Recombinant Mouse Monoclonal Antibody (R35001_89F4)

Product Code. 30828525
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828525 50 μL
30827782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828525 Supplier Invitrogen™ Supplier No. MA564522

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

HOMER1 Recombinant Monoclonal Antibody for IHC (P)

HOMER1 is a member of the homer family of dendritic proteins. Members of this family regulate group 1 metabotrophic glutamate receptor function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOMER1
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone R35001_89F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene Homer1
Gene Accession No. Q86YM7
Gene Alias HOMER; homer homolog 1; homer homolog 1 (Drosophila); homer protein homolog 1; homer scaffolding protein 1; homer, neuronal immediate early gene, 1; Homer1; homer-1; HOMER1A; HOMER1B; HOMER1C; HOMER1F; immediate early gene protein Homer1A; PSD-Zip45; scaffold protein; SYN47; VASP/Ena-related gene up-regulated during seizure and LTP 1; Ves-1; Vesl; Vesl1; Vesl-1
Gene Symbols Homer1
Host Species Mouse
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9456
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.