missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOMER1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12433-1-AP-20UL
This item is not returnable.
View return policy
Description
HOMER1 is a member of the homer family of dendritic proteins. Members of this family regulate group 1 metabotrophic glutamate receptor function.Specifications
| HOMER1 | |
| Polyclonal | |
| Unconjugated | |
| HOMER1 | |
| HOMER, Homer 1, homer homolog 1 (Drosophila), Homer protein homolog 1, HOMER1, HOMER1A, HOMER1B, HOMER1C, SYN47, Ves 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26556, 29546, 9456 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.46 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q86YM7, Q9Z214, Q9Z2Y3 | |
| HOMER1 | |
| HOMER1 Fusion Protein Ag3109 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction