Learn More
HOMER1 Mouse, anti-Human, Clone: 3B1B11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68597-1-IG-20UL
Description
The HOMER1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HOMER1. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The HOMER1 antibody has been validated for the following applications: WB,IF,ELISA analysis.
Specifications
| HOMER1 | |
| 3B1B11 | |
| HOMER, Homer 1, homer homolog 1 (Drosophila), Homer protein homolog 1, HOMER1, HOMER1A, HOMER1B, HOMER1C, SYN47, Ves 1 | |
| 20 μL | |
| 9456 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunofluorescence, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Pig | |
| 150 kDa | |
| IgG1 |