missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HOMER1 Monoclonal Antibody (OTI7G10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527522
This item is not returnable.
View return policy
Description
HOMER1 Monoclonal Antibody for Western Blot, IHC (P)
HOMER1 is a member of the homer family of dendritic proteins. Members of this family regulate group 1 metabotrophic glutamate receptor function.
Specifications
| HOMER1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86YM7, Q9Z214, Q9Z2Y3 | |
| Homer1 | |
| Human recombinant protein fragment corresponding to amino acids 1-354 of HOMER1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7G10 | |
| Unconjugated | |
| Homer1 | |
| HOMER; homer homolog 1; homer homolog 1 (Drosophila); homer protein homolog 1; homer scaffolding protein 1; homer, neuronal immediate early gene, 1; Homer1; homer-1; HOMER1A; HOMER1B; HOMER1C; HOMER1F; immediate early gene protein Homer1A; PSD-Zip45; scaffold protein; SYN47; VASP/Ena-related gene up-regulated during seizure and LTP 1; Ves-1; Vesl; Vesl1; Vesl-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 26556, 29546, 9456 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction