missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homeodomain Interacting Protein Kinase 3 Rabbit Anti-Human, Polyclonal, Abnova&Trade;
Rabbit Polyclonal Antibody
Brand: Abnova PAB2938.400uL
This item is not returnable.
View return policy
Description
homeodomain interacting protein kinase 3 Polyclonal antibody specifically detects homeodomain interacting protein kinase 3 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| homeodomain interacting protein kinase 3 | |
| Polyclonal | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. |
|
| DYRK6/FIST3/PKY/YAK1 | |
| A mixture of synthetic peptide (conjugated with KLH) corresponding to residues within 1100-1200 and 1134-1163 of human HIPK3. | |
| RUO | |
| 10114 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| In PBS (0.09% sodium azide) | |
| Rabbit | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction