missing translation for 'onlineSavingsMsg'
Learn More

Homatropine Methylbromide, TRC

Product Code. 30392134
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392134 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392134 Supplier Toronto Research Chemicals Supplier No. TRCH459225100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Homatropine methylbromide
CAS 80-49-9
Molecular Formula C17 H24 N O3 . Br
Quantity 100 g
Synonym Homatropine Methylbromide, (1R,3r,5S)-3-[[(2RS)-2-Hydroxy-2-phenylacetyl]oxy]-8,8-dimethyl-8-azoniabicyclo[3.2.1]octane bromide
InChI Formula InChI=1S/C17H24NO3.BrH/c1-18(2)13-8-9-14(18)11-15(10-13)21-17(20)16(19)12-6-4-3-5-7-12;/h3-7,13-16,19H,8-11H2,1-2H3;1H/q+1;/p-1/t13-,14+,15+,16-;/m1./s1
SMILES [Br-].C[N+]1(C)[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@H](O)c3ccccc3
IUPAC Name [(1S,5R)-8,8-dimethyl-8-azoniabicyclo[3.2.1]octan-3-yl] (2R)-2-hydroxy-2-phenylacetate;bromide
Molecular Weight (g/mol) 370.28
Formula Weight 369.094
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.