missing translation for 'onlineSavingsMsg'
Learn More

Homarine, TRC

Product Code. 30405152
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30405152 50 mg
30402475 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30405152 Supplier Toronto Research Chemicals Supplier No. TRCH45850050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 445-30-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
Synonym 2-Carboxy-1-methylpyridinium Inner Salt, 2-Pyridinecarboxylic Acid N-Methylbetaine, 2-Pyridinecarboxylic Acid Methoinner Salt
SMILES Cl.NC(=N)NCCC[C@H](N)C(O)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H7NO2/c1-8-5-3-2-4-6(8)7(9)10/h2-5H,1H3
Formula Weight 137.0477
Percent Purity >95%
Chemical Name or Material Homarine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.