missing translation for 'onlineSavingsMsg'
Learn More

Holomycin, TRC

Product Code. 30400959
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30400959 2.5 mg 2.5mg
30397426 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400959 Supplier Toronto Research Chemicals Supplier No. TRCH4584902.5MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-04-0
Molecular Formula C7 H6 N2 O2 S2
Molecular Weight (g/mol) 214.26
Synonym N-(4,5-Dihydro-5-oxo-1,2-dithiolo[4,3-b]pyrrol-6-yl)acetamide, 6-Acetamido-1,2-dithiolo[4,3-b]pyrrol-5(4H)-one, 6-(Acetylamino)-1,2-dithiolo[4,3-b]pyrrol-5(4H)-one
IUPAC Name N-(5-oxo-4H-dithiolo[4,3-b]pyrrol-6-yl)acetamide
SMILES CC(=O)NC1=C2SSC=C2NC1=O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C7H6N2O2S2/c1-3(10)8-5-6-4(2-12-13-6)9-7(5)11/h2H,1H3,(H,8,10)(H,9,11)
Formula Weight 213.9871
Percent Purity >95%
Chemical Name or Material Holomycin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.