missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) trifluoromethanesulfonate hydrate, 98%, Thermo Scientific Chemicals

Product Code. 11393858
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393858 50 g 50g
11303868 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393858 Supplier Thermo Scientific Alfa Aesar Supplier No. 040307.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139177-63-2
Molecular Formula C3F9HoO9S3
Molecular Weight (g/mol) 612.12
MDL Number MFCD00209590
InChI Key DBPDCYACEYLEMY-UHFFFAOYSA-K
Synonym holmium iii trifluoromethanesulfonate, holmium triflate, holmium iii trifluoromethanesulphonate, trifluoromethanesulfonic acid holmium iii salt, methanesulfonic acid,1,1,1-trifluoro-, holmium 3+ salt 3:1, holmium trifluoromethanesulfonate, holmium iii triflate, ho otf 3, acmc-1bwo7
PubChem CID 22482917
IUPAC Name holmium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Ho+3]

Specifications

Physical Form Powder
Quantity 50 g
Formula Weight 612.13 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Holmium(III) trifluoromethanesulfonate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.