missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) fluoride, anhydrous, 99.9% (REO)

Product Code. 15452107
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452107 50 g 50g
15432107 2 g 2g
15442107 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15452107 Supplier Thermo Scientific Alfa Aesar Supplier No. 045033.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13760-78-6 | F3Ho | 221.93 g/mol

Holmium(III) fluoride, anhydrous is used to produce the strongest artificially- generated magnetic fields, nuclear control rods, yellow-colored glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13760-78-6
Molecular Formula F3Ho
Molecular Weight (g/mol) 221.93
MDL Number MFCD00016142
InChI Key FDIFPFNHNADKFC-UHFFFAOYSA-K
IUPAC Name holmium(3+) trifluoride
SMILES [F-].[F-].[F-].[Ho+3]

Specifications

Melting Point 1143°C
Physical Form Powder
Quantity 50 g
UN Number UN3288
Solubility Information Insoluble in water.
Formula Weight 221.93
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 7.6 g/mL
Assay (REO)
Chemical Name or Material Holmium (III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.