missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) fluoride, anhydrous, 99.9% (REO)

Product Code. 11460391
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460391 2 g 2g
11334118 10 g 10g
11324118 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460391 Supplier Thermo Scientific Alfa Aesar Supplier No. 041413.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13760-78-6 | F3Ho | 221.93 g/mol

For vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13760-78-6
Molecular Formula F3Ho
Molecular Weight (g/mol) 221.93
MDL Number MFCD00016142
InChI Key FDIFPFNHNADKFC-UHFFFAOYSA-K
IUPAC Name holmium(3+) trifluoride
SMILES [F-].[F-].[F-].[Ho+3]

Specifications

Melting Point 1143°C
Physical Form Pieces
Quantity 2 g
UN Number UN3288
Formula Weight 221.93
Percent Purity 99.9%
Packaging Glass bottle
Sensitivity Hygroscopic
Specific Gravity 7.6
Density 7.6
Chemical Name or Material Holmium(III) fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.