missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Holmium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
92.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
Holmium(III) chloride is used in nuclear control rods, microwaves, cubic zirconia, coloring glass yellow, solid state lasers and magnetic poles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 718°C |
Physical Form | Crystalline Aggregates |
Quantity | 10 g |
Merck Index | 14,4727 |
Solubility Information | Soluble in water. |
Formula Weight | 271.29 (Anhydrous) |
Percent Purity | 99.99% |
Grade | Reacton™ |
Sensitivity | Hygroscopic |
Assay | (REO) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'