missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Holmium(III) chloride hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Holmium(III) chloride hexahydrate is used as laboratory reagent and also used in proteomics research , optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 718°C |
Physical Form | Crystalline Aggregates |
Quantity | 100 g |
Linear Formula | HoCl3·6H2O |
Merck Index | 14,4727 |
Solubility Information | Soluble in water. |
IUPAC Name | trichloroholmium hexahydrate |
Formula Weight | 379.38 (271.29 Anhydrous) |
Percent Purity | 99.9% |
Grade | Reacton™ |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'