missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) bromide hydrate, REacton™, 99.99% (REO)

Product Code. 11366648
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366648 50 g 50g
11376648 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11366648 Supplier Thermo Scientific Alfa Aesar Supplier No. 013203.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 223911-98-6 | Br3Ho | 404.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 223911-98-6
Molecular Formula Br3Ho
Molecular Weight (g/mol) 404.64
MDL Number MFCD00209652
InChI Key MZNSYJWLQLXLHE-UHFFFAOYSA-K
Synonym holmium iii bromide hydrate, holmium bromide hydrate, holmium bromide hobr3 , hydrate 9ci
PubChem CID 57375929
SMILES [Br-].[Br-].[Br-].[Ho+3]

Specifications

Melting Point 914°C
Physical Form Crystalline
Quantity 50 g
Merck Index 14,4727
IUPAC Name holmium(3+);tribromide;hydrate
Formula Weight 404.66 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Holmium(III) bromide hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.