missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog Number 11376648
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
223911-98-6
Br3Ho
404.64
MFCD00209652
MZNSYJWLQLXLHE-UHFFFAOYSA-K
holmium iii bromide hydrate, holmium bromide hydrate, holmium bromide hobr3 , hydrate 9ci
57375929
[Br-].[Br-].[Br-].[Ho+3]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
223911-98-6
Br3Ho
404.64
MFCD00209652
MZNSYJWLQLXLHE-UHFFFAOYSA-K
holmium iii bromide hydrate, holmium bromide hydrate, holmium bromide hobr3 , hydrate 9ci
57375929
[Br-].[Br-].[Br-].[Ho+3]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 914°C
Physical Form Crystalline
Quantity 10 g
Merck Index 14,4727
IUPAC Name holmium(3+) tribromide
Formula Weight 404.66 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Holmium(III) bromide hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'