missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) acetate hydrate, REacton™, 99.9% (REO)

Product Code. 11450470
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450470 2 g 2g
11362538 10 g 10g
11372538 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450470 Supplier Thermo Scientific Alfa Aesar Supplier No. 014591.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 312619-49-1 | C6H9HoO6 | 342.06 g/mol

Holmium(III) acetate hydrate is used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312619-49-1
Molecular Formula C6H9HoO6
Molecular Weight (g/mol) 342.06
MDL Number MFCD00150648
InChI Key HWQRVFRWLQGBFV-UHFFFAOYSA-K
Synonym holmium iii acetate hexahydrate, holmium iii acetate hydrate, reacton reo, holmium iii acetatehexahydrate, holmium acetate-water 1/3/6
IUPAC Name holmium(3+) triacetate
SMILES [Ho+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 2 g
Solubility Information Soluble in water.
Formula Weight 342.07 (Anhydrous)
Percent Purity 99.9%
Grade Reacton
Assay (REO)
Chemical Name or Material Holmium(III) acetate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.