Learn More
Thermo Scientific™ Hollow Cathode Single-Element Lamps - Uncoded
Product Code. 17179548 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Hollow Cathode Lamps, Uncoded
Lamp:
Rubidium Uncoded
Product Code. Type Lamp
17179548 Hollow Cathode Lamps, Uncoded Rubidium Uncoded
1 options
Product Code. 17179548 Supplier Thermo Scientific™ Supplier No. 942339020371
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
For maximum flexibility from your Thermo Scientific™ iCE™ 3000 Series AAS system, we offer a huge range of pre-coded and un-coded Hollow Cathode Lamps.
For maximum flexibility from your Thermo Scientific™ iCE™ 3000 Series Atomic Absorption spectrometer, a huge range of pre-coded and un-coded Thermo Scientific are available, to handle your chosen elements for analysis. Whether you require a single dedicated element or a multi-element cathode lamp, there is a one to match your needs, whether coded or un-coded.
For maximum flexibility from your Thermo Scientific™ iCE™ 3000 Series Atomic Absorption spectrometer, a huge range of pre-coded and un-coded Thermo Scientific are available, to handle your chosen elements for analysis. Whether you require a single dedicated element or a multi-element cathode lamp, there is a one to match your needs, whether coded or un-coded.
Specifications
For Use With (Equipment) iCE 3000 Series Atomic Absorption Spectrometers
Type Hollow Cathode Lamps, Uncoded
Lamp Rubidium Uncoded