missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Holder Removing Aid for Buckets 30553104
Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
Bucket 30553104
Unit Size:
Each
Description
- Holder Removing Aid for Buckets
- Compatible with Centrifuge Multi FC5916R 120V and Centrifuge Multi FC5916 120V
Specifications
Specifications
| For Use With (Equipment) | Bucket 30553104 |
| Color | Silver |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction