missing translation for 'onlineSavingsMsg'
Learn More

HOKU-81, MedChemExpress

Product Code. 16443756
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16443756 25 mg 25mg
16420538 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16443756 Supplier Medchem Express Supplier No. HY50291
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress HOKU-81 (4-Hydroxytulobuterol) is one of the metabolites of Tulobuterol (HY-B1810). HOKU-81 is a potent and selective β2-adrenoceptor stimulant. HOKU-81 has bronchodilating effect.

HOKU-81 (4-Hydroxytulobuterol) is one of the metabolites of Tulobuterol (HY-B1810). HOKU-81 is a potent and selective β2-adrenoceptor stimulant. HOKU-81 has bronchodilating effect.

Specifications

Chemical Name or Material HOKU-81
CAS 58020-43-2
Color White
Purity Grade Notes Research
Molecular Formula C12H18ClNO2
Quantity 25 mg
Synonym 4-Hydroxytulobuterol
Solubility Information DMSO : 50 mg/mL (205.15 mM; Need ultrasonic)
SMILES ClC1=C(C(O)CNC(C)(C)C)C=CC(O)=C1
Molecular Weight (g/mol) 243.73
Formula Weight 243.73
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.