missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOIP/RNF31 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48914-25ul
This item is not returnable.
View return policy
Description
HOIP/RNF31 Polyclonal antibody specifically detects HOIP/RNF31 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| HOIP/RNF31 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| EC 6.3.2.-, FLJ10111, FLJ23501, HOIP, MGC19975, ring finger protein 31, ZIBRAHOIL-1-interacting protein, Zinc in-between-RING-finger ubiquitin-associated domain protein | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ELALSLLQETPRNYELGDVVEAVRHSQDRAFLRRLLAQECAVCGWALPHNRMQALTSCECTICPDCFRQHFTIALKEKHITDMVCPACGRPDLTDDTQLLSYFSTLDIQLRESLEPDAYALFHKKLTEGV | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55072 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction