missing translation for 'onlineSavingsMsg'
Learn More

HOIP/RNF31 Antibody [DyLight 680], Novus Biologicals™

Product Code. 30734029 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734029 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734029 Supplier Novus Biologicals Supplier No. AF8039Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

HOIP/RNF31 Polyclonal antibody specifically detects HOIP/RNF31 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOIP/RNF31
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2.-, FLJ10111, FLJ23501, HOIP, MGC19975, ring finger protein 31, ZIBRAHOIL-1-interacting protein, Zinc in-between-RING-finger ubiquitin-associated domain protein
Host Species Sheep
Immunogen E. coli-derived recombinant human HOIP/RNF31, Arg970-Lys1072, Accession # Q96EP0
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55072
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.