missing translation for 'onlineSavingsMsg'
Learn More

HOGA1 Antibody, HRP, Novus Biologicals™

Product Code. 18368912 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368912 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368912 Supplier Novus Biologicals Supplier No. NBP306562H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HOGA1 Polyclonal antibody specifically detects HOGA1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOGA1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias 4-hydroxy-2-oxoglutarate aldolase 1, C10orf65, chromosome 10 open reading frame 65, DHDPS2, DHDPSLHP3, DHDPS-like protein, Dihydrodipicolinate synthase-like, dihydrodipicolinate synthase-like, mitochondrial, dihydrodipicolinate synthetase homolog 2, EC 4.1.3.16, FLJ37472, FLJ56960, N-acetylneuraminate pyruvate lyase 2 (putative), NPL2, Probable 2-keto-4-hydroxyglutarate aldolase, probable 4-hydroxy-2-oxoglutarate aldolase, mitochondrial, Probable KHG-aldolase, Protein 569272
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human HOGA1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112817
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.