missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ TE70X and TE77X Semidry Blotting Systems
Click to view available options
Type:
Semi Dry Electroblotting System
Includes:
21 x 26 cm Blot Paper
Built-in power supply, Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (21 x 26cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™
Power Supply, 14 x 16 cm Blot Paper
Unit Size:
Each
Description
Includes
Molded base with iridium oxide anode, top assembly with stainless-steel cathode, power cables, 25 sheets of blotting paper, 50 sheets of cellophane (Mfr. Nos. TE70X and TE70XP only), two solid Mylar™ masks
- Transfer up to two polyacrylamide gels in a stack
- Use minimal buffer
- Do not generate the excessive heat that can distort bands or denature protein
- Utilize durable iridium oxide and stainless-steel electrodes to assure uniform, contamination-free transfers
- Offer automatic transfer-stopping feature to prevent overheating when buffer is depleted
- Available with built-in power supply to save space and simplify transfer
Certifications
CE marked, CSA C22.2 1010.1, EN61010-1, UL3101-1
Specifications
Specifications
| Certifications/Compliance | EN61010-1, UL3101-1, CSA C22.2 1010.1, CE |
| Content And Storage | Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (21 x 26cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™ masks (16.5 x |
| For Use With (Equipment) | Electrophoresis Blotting System |
| Type | Semi Dry Electroblotting System |
| Humidity Range | Up to 80% |
| Includes | 21 x 26 cm Blot Paper |
| Temperature (English) Maximum | 113°F |
| Temperature (Metric) Maximum | 45°C |
| Dimensions (L x W x H) | 18.89 x 15 x 4 in. (48 x 38 x 10 cm) |
| Length (English) | 18.89 in. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
