missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Sponges

Product Code. 12074637
Change view
Click to view available options
For Use With (Equipment):
TE 22 Mini Tank Transfer Unit
TE 42, TE 62 Transfer Tank
Material:
Dacron
Foam
Thickness (English):
0.11 in.
0.12 in.
0.23 in.
0.25 in.
Thickness (Metric):
0.3 cm
0.6 cm
Unit Size:
Each
Pack of 4
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Material Thickness (English) Thickness (Metric) Unit Size
12074637 TE 42, TE 62 Transfer Tank Foam 0.25 in. 0.6 cm Pack of 4
11514444 TE 42, TE 62 Transfer Tank Dacron 0.25 in. 0.6 cm Each
12014727 TE 22 Mini Tank Transfer Unit Foam 0.11 in. 0.3 cm Pack of 4
11525634 TE 42, TE 62 Transfer Tank Foam 0.12 in. 0.3 cm Pack of 4
12069927 TE 42, TE 62 Transfer Tank Foam 0.12 in. 0.3 cm Pack of 4
12029967 TE 22 Mini Tank Transfer Unit Foam 0.23 in. 0.6 cm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 12074637 Supplier Hoefer™ Supplier No. TE45F
  • ONLINE EXCLUSIVE PRICE
    £33.25
    £28.34 / Pack of 4
    Save £4.91
    14% Off
Estimated Shipment: 26-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with TE42 Standard Transfer Tank

  • 3 or 6 mm Thick
  • 4 pcs

Specifications

For Use With (Equipment) TE 42, TE 62 Transfer Tank
Type Sponge
Material Foam
Quantity 4
Thickness (English) 0.25 in.
Thickness (Metric) 0.6 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.