Learn More
Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Electrode Panels
For use with TE 42 and TE 62 Transfer Tank
Brand: Hoefer™ TE23
Code : 04
Additional Details : Weight : 0.80000kg
Description
- Comes in gray or black electrode panels
- Type: Gray Electrode Panel
- Includes Banana plug and platinum wire
Specifications
TE 22 Mini Tank Transfer Unit | |
Electrode Panel | |
Banana plug and Platinum wire |