missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Electrode Panels

Product Code. 10321095 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
TE 22 Mini Tank Transfer Unit
TE 42, TE 62 Transfer Tank
Type:
Black Electrode Panel
Electrode Panel
Gray Electrode Panel
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Type unitSize
10321095 TE 42, TE 62 Transfer Tank Black Electrode Panel Each
12099857 TE 42, TE 62 Transfer Tank Gray Electrode Panel Each
12054747 TE 42, TE 62 Transfer Tank Black Electrode Panel Each
12084817 TE 22 Mini Tank Transfer Unit Electrode Panel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10321095 Supplier Hoefer™ Supplier No. TE43BK

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with TE 42 and TE 62 Transfer Tank

  • Comes in gray or black electrode panels
  • Type: Gray Electrode Panel
  • Includes Banana plug and platinum wire
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) TE 42, TE 62 Transfer Tank
Type Black Electrode Panel
Includes Banana plug and Platinum wire
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.