missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Submarine Gel Electrophoresis Units: SUB20 Maxi-Standard Horizontal

Product Code. 12655766
Click to view available options
:
Maxi-Std. submarine; 47.5L x 27W x 8cmH; Gel: 20L x 20cmW
This item is not returnable. View return policy

Product Code. 12655766

Brand: Hoefer™ SUB20

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for high-resolution analytical and preparative studies of nucleic acids

  • Provides a 20 x 20 cm gel-running tray w/four comb positions
  • Handles up to 168 samples
  • Multi-channel pipette compatible combs with a max. 30 sample throughput reduce gel loading time
  • Buffer recirculation ports
  • Color-coded and height-adjustable combs
  • Colored loading strips
  • Compact tank
  • UV-transparent running tray
  • Side handles for safe, easy transport

Specifications

Certifications/Compliance EN61010-1, CE
For Use With (Application) Nucleic Acid Gel Electrophoresis
Type Maxi Standard Submarine Gel Unit
Gel Dimensions 20 x 20 cm
Humidity Range 80%
Includes Buffer chamber assembly, safety lid with high voltage Leads, running tray with casting gasket, 1mm thick, 16-well combs, colored loading strips, Buffer recirculation ports
Sample Size 160
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Volume (Fill) Metric 2.2 L
Dimensions (L x W) 18.7 x 10.62 in. (47.5 x 27 cm)
Length (English) 18.7 in.
Length (English) Gel 7.87 in.
Length (Metric) 47.5 cm
Length (Metric) Gel 20 cm
Width (English) 10.62 in.
Width (English) Gel 7.87 in.
Width (Metric) 27 cm
Width (Metric) Gel 20 cm
Height (Metric) 8 cm
Voltage 1000 V
Wattage 500 w
Amperage 500 mA
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.