missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Submarine Gel Electrophoresis Units: SUB15 Standard Horizontal
Description
- Highly versatile option for screening nucleic acid samples ranging from DNA mini-preps to PCR products
- 120-sample maximum throughput
- 5 x 15 cm gel tray, with comb options ranging from 1 to 30 samples
- Multi-channel pipette compatible combs with a max. 30 sample throughput reduce gel loading time
- Buffer recirculation ports
- Color-coded and height-adjustable combs
- Colored loading strips
- Compact tank
- UV-transparent running tray
- Side handles for safe, easy transport
Specifications
Specifications
| Certifications/Compliance | EN61010-1, CE |
| For Use With (Application) | Screening samples ranging from DNA mini preparation to PCR product |
| Type | Standard Submarine Gel Electrophoresis Unit |
| Humidity Range | 80% |
| Includes | Buffer chamber assembly, safety lid with high voltage Leads, running tray with Silicone gasket, 1mm thick, 16-well combs, colored loading strips, Buffer recirculation ports |
| Sample Size | 120 |
| Temperature (English) Maximum | 113°F |
| Temperature (Metric) Maximum | 45°C |
| Volume (Fill) Metric | 1.2 L |
| Dimensions (L x W) | 13.18 x 8.46 in. (33.5 x 21.5 cm) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?