missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Submarine Gel Electrophoresis Units: SUB13 Midi Horizontal

Product Code. 12605736
Click to view available options
:
Midi submarine; 32L x 20W x 7cmH; Gel: 15L x 12.8cmW
This item is not returnable. View return policy

Product Code. 12605736

Brand: Hoefer™ SUB13

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for analytical and preparative studies of nucleic acids

  • Maximum 112-sample throughput capacity and features a removable gel-casting tray with end gaskets
  • Buffer recirculation ports
  • Color-coded and height-adjustable combs
  • Colored loading strips
  • Compact tank
  • UV-transparent running tray
  • Side handles for safe, easy transport

Specifications

Certifications/Compliance EN61010-1, CE
For Use With (Application) Nucleic Acid Gel Electrophoresis
Type Midi Submarine Gel Electrophoresis Unit
Gel Dimensions 15 x 12.8 cm
Humidity Range 80%
Includes Buffer chamber assembly, safety lid with high voltage Leads, running tray with Silicone gasket, 1mm thick, 16-well combs, colored loading strips, Buffer recirculation ports
Sample Size 112
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Volume (Fill) Metric 0.9 L
Dimensions (L x W) 12.59 x 7.87 in. (32 x 20 cm)
Length (English) 12.59 in.
Length (English) Gel 5.90 in.
Length (Metric) 32 cm
Length (Metric) Gel 15 cm
Width (English) 7.87 in.
Width (English) Gel 5.03 in.
Width (Metric) 20 cm
Width (Metric) Gel 12.8 cm
Height (Metric) 7 cm
Voltage 300 V
Wattage 60 w
Amperage 200 mA
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.