Learn More
Hoefer™ SUB15 Standard Submarine Combs
Accessories for SUB15 Standard Submarine
Brand:  Hoefer™ SUB15-C1.5-10
 View more versions of this product
Product Code. 12695760
  • £67.39 / Each
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ranges from 10, 16, 20, 25 and 30 Wells
  • 1, 1.5 and 2 mm Thickness
  • 2.5 to 12.8 mm Well Width
  • Min. 12, max. 128 μL Sample Volume
Specifications
Specifications
SUB15 Standard Submarine
10
0.15 cm
0.5 in.
0.19 in.
Comb
0.05 in.
96 μL
1.28 cm
0.5 cm