missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Spring Clamps for Mighty Small™ Electrophoresis Units
Description
- Multiple Gel Casters Replacement Parts
- Some are Asymetrical Clamps
Specifications
Specifications
| Color | Red |
| For Use With (Equipment) | SE 250, SE 260 Mini Vertical Electrophoresis Unit, SE 615, SE 675 Multiple Gel Casters |
| Type | Spring Clamp |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?