missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Spacers for SE 600 Chroma, SE 600 Standard, or SE 400 Vertical Electrophoresis Units

Accessory parts for SE 600 Chroma SE 600 Standard or SE 400 Vertical Electrophoresis Units

Brand:  Hoefer™ SE6118-2-1.5

 View more versions of this product

Product Code. 12034787

  • £65.04 / Pack of 2
Estimated Shipment: 04-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ranges from 0.75 to 3.0 mm Thickness
  • 8, 16 and 24 cm in Length
  • 1 to 2 cm Width
  • 2 Pk.
Specifications

Specifications

SE 600 Chroma, SE 600 Standard, SE 400 Vertical Electrophoresis Unit
1 x 16 cm x 1.5mm
0.05 in.
16 cm
Spacer
2/Pk.
0.15 cm
0.1 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers