Learn More
Hoefer™ Space Saver Plate for SE900 Electrophoresis Units
For use with SE900 Large Format Vertical Gel Protein Electrophoresis Unit
Brand: Hoefer™ SE912
Code : 04
Additional Details : Weight : 0.10000kg
Description
- Accessories and Replacement Parts for SE900 Units
Specifications
Space Saver Plate | |
SE900-1.0 and SE900 Electrophoresis Units |