missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ SG Series Gradient Makers

Product Code. 11570764 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
15 mL
30 mL
50 mL
100 mL
500 mL
Unit Size:
15mL
500mL
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11570764 50 mL Each
10089259 100 mL Each
11564424 100 mL Each
12089937 50 mL Each
11544424 30 mL Each
12074647 500 mL Each
10049309 500 mL 500mL
11534424 15 mL Each
10714762 15 mL 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 11570764 Supplier Hoefer™ Supplier No. SG50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

  • Models SG 15, SG 30, SG 50, and SG 100 include gradient maker with PTFE valves, Luer outlet fitting, support rod, adapter barbed fitting, 22-gauge needle.
  • Model SG 500 includes gradient maker with push-pull valve, four wrap-and-seal hose clamps.

Ideal for generating gradients of polyacrylamide, sucrose, and cesium chloride

Models SG 15, SG 30, SG 50, and SG 100

  • Milled of heavy acrylic plastic with high-quality, leak-free PTFE-resin valves
  • Offer a flat base for stability on magnetic stirrers; support rod can be attached to ring stand for additional stability
  • Accept Luer or 4 mm ID tubing; with included adapters can also accept 2 mm ID or 22-gauge tubing
  • SG 100 is ideal for the Hoefer Mighty Small™ Gel Casters SE 275 (for four gels) or SE 215 (for 10 gels)
Model SG 500
  • Made of two cylindrical acrylic chambers joined and mounted on a flat acrylic base
  • Uses a simple push-pull valve to open and close the passage between the reservoir and mixing chambers
  • Suitable for forming polyacrylamide gradients or buffer gradients used with medium to small chromatography columns
  • Ideal for use with the Hoefer SE 615 or SE 915 Multiple Gel Casters
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mighty Small™ Gel Caster SE 275 (for 4 Gels) or SE 215 (for 10 Gels)
Type Gradient Maker
Includes Gradient maker w/PTFE Valves , Luer outlet fitting, support rod, adapter barbed fitting and 22G needle
Volume (English) 1.69 oz.
Volume (Metric) 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.