Learn More
Hoefer™ SG Gradient Maker: Stopcock Valve
Stopcock valve for SG Gradient Makers
Brand: Hoefer™ SG100-1
Code : NEW
Additional Details : Weight : 0.80000kg
Description
For use with SG Gradient Makers
Specifications
SG 5, SG 15, SG 30 or SG 50 Gradient Maker | |
Stopcock Valve | |
PTFE |