Learn More
Hoefer™ SE660 Tall Standard Dual Cooled Vertical Unit
Description
- Dual-Cooled
Specifications
Specifications
| Certifications/Compliance | EN61010-1, UL61010-1, CSA C22.2 1010.1, CE |
| Humidity Range | Up to 80% |
| Temperature (Metric) Maximum | 45°C |
| Depth (Metric) | 14 cm |