Learn More
Hoefer™ SE6160 Heat exchanger accessories
Product Code. 15568991
Change view
Click to view available options
Type:
Grommet
Tube and Grommet
Unit Size:
Pack of 4
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
15568991 Tube and Grommet Pack of 5
15518991 Grommet Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15568991 Supplier Hoefer™ Supplier No. SE61605
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ergonomic design offers easier handling and sample introduction.
  • Basic Unit
  • Accessories
Specifications
For Use With (Equipment) SE 6160 Heat Exchanger/Lower Electrode Assembly
Type Tube and Grommet