Learn More
Cytiva Accessories for SE 250/260 Mini-vertical Gel Units: Alumina Notched Plate
Product Code. 10279714 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1/Pk.
10/Pk.
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10279714 1/Pk. Each
11735393 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10279714 Supplier Cytiva Supplier No. SE202N
Only null left
Add to basket
Add to basket
Specifications
For Use With (Equipment) SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit
Type Notched Alumina Plate
Material Alumina
Quantity 1/Pk.
Length (Metric) 10 cm
Width (Metric) 8 cm