missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Maxi-Plus and Maxi-Plus Submarine Gel Electrophoresis Units

Product Code. 12665766
Change view
Click to view available options
Length (English) Gel:
7.8 in.
9.8 in.
11.8 in.
Width (English) Gel:
7.8 in.
9.8 in.
11.8 in.
Humidity Range:
80%
Up to 80%
Sample Size:
160
624
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Gel Width (English) Gel Humidity Range Sample Size Unit Size
12665766 7.8 in. 7.8 in. Up to 80% 160 Each
12666203 9.8 in. 11.8 in. 80% 624 Each
12665786 11.8 in. 9.8 in. Up to 80% 624 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12665766 Supplier Hoefer™ Supplier No. SUB20C
  • ONLINE EXCLUSIVE PRICE
    £1055.00
    £857.10 / Pack of 1
    Save £197.90
    18% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for high resolution analytical and preparative studies of nucleic acids

  • Large format ideal for high resolution techniques
  • Multichannel pipette compatible combs
  • Color-coded and height adjustable combs
  • Buffer recirculation ports
  • Colored loading strips
  • UV-transparent running tray
  • Side handles for safe, easy transport

Specifications

Certifications/Compliance EN61010-1, CE
Type Submarine Gel Electrophoresis Unit
Humidity Range Up to 80%
Includes Buffer chamber assembly with cooling ports, safety lid with high voltage Leads, running tray with casting gates, 1mm thick, 16-well combs, colored loading strips, Buffer recirculation ports
Sample Size 160
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Volume (English) Buffer 74.3 oz.
Volume (Metric) Buffer 2200 mL
Dimensions (L x W x H) 18.7 x 10.6 x 3.1 in. (47.5 x 27 x 8 cm)
Length (English) 18.7 in.
Length (English) Gel 7.8 in.
Length (Metric) 47.5 cm
Length (Metric) Gel 20 cm
Width (English) 10.6 in.
Width (English) Gel 7.8 in.
Width (Metric) 27 cm
Width (Metric) Gel 20 cm
Height (English) 3.1 in.
Height (Metric) 8 cm
Voltage 1000 V
Wattage 500 w
Amperage 500 mA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.